1,3-Benzothiazole-6-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 6-Benzothiazolecarboxaldehyde, Benzo[d]thiazole-6-carbaldehyde, Benzothiazol-6-carboxaldehyde, 6-Formylbenzothiazole

A156482-1G A156482-250MG A156482-5G A156482-100MG A156482-25G
76973-328EA 69 USD
76973-328 76973-330 76973-332 77671-204 77881-226
1,3-Benzothiazole-6-carboxaldehyde 97%
1,3-Benzothiazole-6-carboxaldehyde

Precaución: for Research Use Only

Formula: C₈H₅NOS
MDL Number: MFCD03086101
CAS Number: 19989-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR