Rilpivirine 98%

Supplier: Ambeed
Warning

A112484-100MG A112484-1G A112484-1MG A112484-50MG A112484-5MG A112484-250MG
76970-904EA 79 USD
76970-904 76970-906 76970-908 76970-910 76970-912 76970-914 77570-688
Rilpivirine 98%
Rilpivirine

Precaución: for Research Use Only

Formula: C₂₂H₁₈N₆
MDL Number: MFCD11046372
CAS Number: 500287-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR