2-Amino-5-bromo-4-isopropylthiazole 96%

Supplier: Ambeed
Warning

A666795-100MG A666795-1G A666795-250MG
76980-902EA 35.01 USD
76980-902 76980-904 76980-906
2-Amino-5-bromo-4-isopropylthiazole 96%
2-Amino-5-bromo-4-isopropylthiazole

Precaución: for Research Use Only

Formula: C₆H₉BrN₂S
MW: 221.12 g/mol
MDL Number: MFCD09834790
CAS Number: 1025700-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR