O-Benzyl-L-tyrosine benzyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Tyr(Bzl)-OBzl·HCl, H-Tyr(Bzl)-OBzl.HCl

A635044-5G A635044-25G A635044-1G
76980-698EA 59 USD
76980-698 77882-470 77882-469
O-Benzyl-L-tyrosine benzyl ester hydrochloride 98%
O-Benzyl-L-tyrosine benzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₂₃NO₃·HCl
MDL Number: MFCD00153465
CAS Number: 52142-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR