5-Chloropyrimidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A248403-1G A248403-5G
76976-632EA 70 USD
76976-632 77881-657
5-Chloropyrimidine-2-carboxylic acid 97%
5-Chloropyrimidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₂
MDL Number: MFCD07440094
CAS Number: 38275-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR