2-Bromo-5-formylbenzonitrile 96%

Supplier: Ambeed
Warning

A125033-250MG A125033-1G
76971-802EA 49 USD
76971-802 77571-496
2-Bromo-5-formylbenzonitrile 96%
2-Bromo-5-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrNO
MW: 210.027 g/mol
MDL Number: MFCD18073530
CAS Number: 1228829-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR