1-Isopropenyl-2-benzimidazolidinone 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Prop-1-en-2-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one, 1-(Prop-1-en-2-yl)-1H-benzo[d]imidazol-2(3H)-one

A440024-1G A440024-5G
76979-364EA 5.31 USD
76979-364 76979-366
1-Isopropenyl-2-benzimidazolidinone 98%
1-Isopropenyl-2-benzimidazolidinone

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MW: 174.203 g/mol
MDL Number: MFCD00218253
CAS Number: 52099-72-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR