(1S,3S,5S)-tert-Butyl 3-carbamoyl-2-azabicyclo[3.1.0]hexane-2-carboxylate 97%

Supplier: Ambeed
Warning

A386730-10G A386730-1G A386730-250MG A386730-5G A386730-25G
76978-882EA 136.86 USD
76978-882 76978-884 76978-886 76978-888 77568-792
(1S,3S,5S)-tert-Butyl 3-carbamoyl-2-azabicyclo[3.1.0]hexane-2-carboxylate 97%
(1S,3S,5S)-tert-Butyl 3-carbamoyl-2-azabicyclo[3.1.0]hexane-2-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₈N₂O₃
MW: 226.27 g/mol
MDL Number: MFCD13151933
CAS Number: 361440-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR