3-Bromo-4-(trifluoromethyl)benzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: Benzenemethanol, 3-bromo-4-(trifluoromethyl)

A372637-1G A372637-250MG A372637-5G
76978-722EA 33 USD
76978-722 76978-724 76978-726
3-Bromo-4-(trifluoromethyl)benzyl alcohol 95%
3-Bromo-4-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MDL Number: MFCD13185397
CAS Number: 372120-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR