(S)-Pyrrolidin-2-ylmethanamine dihydrochloride 97%

Supplier: Ambeed
Warning

A112769-250MG A112769-1G A112769-5G
76970-950EA 26 USD
76970-950 77880-908 77880-909
(S)-Pyrrolidin-2-ylmethanamine dihydrochloride 97%
(S)-Pyrrolidin-2-ylmethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄Cl₂N₂
MW: 173.08 g/mol
MDL Number: MFCD11974898
CAS Number: 103382-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR