1-(4-Bromophenyl)-2-phenylbenzimidazole 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromophenyl)-2-phenyl-1H-benzo[d]imidazole

A306280-10G A306280-1G A306280-25G A306280-5G
76977-806EA 79.57 USD
76977-806 76977-808 76977-810 76977-812
1-(4-Bromophenyl)-2-phenylbenzimidazole 95%
1-(4-Bromophenyl)-2-phenylbenzimidazole

Precaución: for Research Use Only

Formula: C₁₉H₁₃BrN₂
MW: 349.23 g/mol
MDL Number: MFCD09746342
CAS Number: 760212-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR