Methyl 2-(cyclopentylamino)acetate hydrochloride 95%

Supplier: Ambeed
Warning

A139039-10G A139039-1G A139039-250MG A139039-5G
76972-510EA 252.5 USD
76972-510 76972-512 76972-514 76972-516
Methyl 2-(cyclopentylamino)acetate hydrochloride 95%
Methyl 2-(cyclopentylamino)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂·ClH
MW: 193.671 g/mol
MDL Number: MFCD15071418
CAS Number: 195877-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR