Ethyl-4-iodobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester
A104892-100G
A104892-10G
A104892-25G
A104892-500G
A104892-5G
76970-354EA
41.38
USD
76970-354
76970-356
76970-358
76970-360
76970-362
Ethyl-4-iodobenzoate 98%
Ethyl-4-iodobenzoate
Precaución: for Research Use Only
Formula:
C₉H₉IO₂ MW: 276.07 g/mol |
MDL Number:
MFCD00017344 CAS Number: 51934-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...