2-Chloro-5-ethylpyridine 96%

Supplier: Ambeed
Warning

A222387-1G A222387-250MG
76975-860EA 304.48 USD
76975-860 76975-862
2-Chloro-5-ethylpyridine 96%
2-Chloro-5-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.59 g/mol
MDL Number: MFCD18384612
CAS Number: 90196-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR