2-Isopropoxypyridine 98%

Supplier: Ambeed
Warning

A307450-1G A307450-250MG
76977-828EA 34 USD
76977-828 76977-830
2-Isopropoxypyridine 98%
2-Isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD11113404
CAS Number: 16096-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR