Methyl 2-bromothiophene-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A159771-1G
A159771-250MG
A159771-25G
A159771-5G
76973-498EA
27
USD
76973-498
76973-500
76973-502
77881-250
Methyl 2-bromothiophene-3-carboxylate 97%
Methyl 2-bromothiophene-3-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₅BrO₂S |
MDL Number:
MFCD16657906 CAS Number: 76360-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...