Hexadecanal 98%

Supplier: Ambeed
Warning

A323037-1G A323037-250MG A323037-5G A323037-100MG A323037-25G
76978-088EA 60.48 USD
76978-088 76978-090 76978-092 77612-866 77672-652
Hexadecanal 98%
Hexadecanal

Precaución: for Research Use Only

Formula: C₁₆H₃₂O
MDL Number: MFCD00055568
CAS Number: 629-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR