Methyl-6-bromo-1H-imidazo[1,2-a]pyridine-8-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-bromoimidazo[1,2-a]pyridine-8-carboxylate

A114037-1G A114037-250MG
76971-056EA 87 USD
76971-056 76971-058
Methyl-6-bromo-1H-imidazo[1,2-a]pyridine-8-carboxylate 95%
Methyl-6-bromo-1H-imidazo[1,2-a]pyridine-8-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MW: 255.08 g/mol
MDL Number: MFCD13195317
CAS Number: 908581-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR