1,8-Dichloro-9,10-bis(phenylethynyl)anthracene 97%

Supplier: Ambeed
Warning

A301336-1G A301336-250MG A301336-25G A301336-5G
76977-730EA 41 USD
76977-730 76977-732 76977-734 76977-736
1,8-Dichloro-9,10-bis(phenylethynyl)anthracene 97%
1,8-Dichloro-9,10-bis(phenylethynyl)anthracene

Precaución: for Research Use Only

Formula: C₃₀H₁₆Cl₂
MDL Number: MFCD00012048
CAS Number: 51749-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR