6-(Hydroxymethyl)pyridine-2-carboxamide 95+%

Supplier: Ambeed
Warning

Synonyms: 6-(Hydroxymethyl)picolinamide

A256569-1G A256569-5G
76976-840EA 31.83 USD
76976-840 76976-842
6-(Hydroxymethyl)pyridine-2-carboxamide 95+%
6-(Hydroxymethyl)pyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD09991807
CAS Number: 41337-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR