N-Methyl-L-proline 97%

Supplier: Ambeed
Warning

Synonyms: N-Me-Pro-OH, H-N-Me-Pro-OH

A163018-10G A163018-1G A163018-250MG A163018-5G A163018-25G
76973-646EA 180.36 USD
76973-646 76973-648 76973-650 76973-652 77671-300
N-Methyl-L-proline 97%
N-Methyl-L-proline

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.159 g/mol
MDL Number: MFCD00011565
CAS Number: 475-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR