4-Hydroxy-2-methylpyrimidine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A110380-100MG A110380-1G A110380-250MG
76970-718EA 9 USD
76970-718 76970-720 76970-722
4-Hydroxy-2-methylpyrimidine-5-carbonitrile 95%
4-Hydroxy-2-methylpyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₅N₃O
MDL Number: MFCD17018685
CAS Number: 27058-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR