5-Bromo-4-methylnicotinaldehyde 98%

Supplier: Ambeed
Warning

A254324-100MG A254324-1G A254324-250MG
76976-750EA 36 USD
76976-750 76976-752 76976-754
5-Bromo-4-methylnicotinaldehyde 98%
5-Bromo-4-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆BrNO
MDL Number: MFCD09909851
CAS Number: 351457-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR