7-Bromo-2,3-dihydro-1,8-naphthyridin-4(1H)-one 97%

Supplier: Ambeed
Warning

A683488-250MG A683488-1G
76981-014EA 204.76 USD
76981-014 77614-114
7-Bromo-2,3-dihydro-1,8-naphthyridin-4(1H)-one 97%
7-Bromo-2,3-dihydro-1,8-naphthyridin-4(1H)-one

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.05 g/mol
MDL Number: MFCD09834136
CAS Number: 64942-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR