5-Bromo-2-methylquinoline 95%

Supplier: Ambeed
Warning

A740016-100MG A740016-1G A740016-250MG A740016-5G
76981-320EA 44.56 USD
76981-320 76981-322 76981-324 76981-326
5-Bromo-2-methylquinoline 95%
5-Bromo-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.081 g/mol
MDL Number: MFCD13189490
CAS Number: 54408-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR