7-Bromobenzo[d]oxazol-2(3H)-one 95%

Supplier: Ambeed
Warning

A150964-100MG A150964-1G A150964-250MG A150964-5G
76973-098EA 43.5 USD
76973-098 76973-100 76973-102 76973-104
7-Bromobenzo[d]oxazol-2(3H)-one 95%
7-Bromobenzo[d]oxazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₄BrNO₂
MW: 214.01 g/mol
MDL Number: MFCD11111731
CAS Number: 871367-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR