7-Bromooxindole 97%

Supplier: Ambeed
Warning

A308252-10G A308252-1G A308252-250MG A308252-25G A308252-5G
76977-836EA 80 USD
76977-836 76977-838 76977-840 76977-842 76977-844
7-Bromooxindole 97%
7-Bromooxindole

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MDL Number: MFCD02179613
CAS Number: 320734-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR