L-4-Methoxymandelic acid 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Hydroxy-2-(4-methoxyphenyl)acetic acid

A123498-10G A123498-1G A123498-250MG A123498-5G
76971-682EA 302.36 USD
76971-682 76971-684 76971-686 76971-688
L-4-Methoxymandelic acid 97%
L-4-Methoxymandelic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.173 g/mol
MDL Number: MFCD00754498
CAS Number: 75172-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR