N-Benzyl-N-Cbz-β-alanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Z-β-N-Cbz-Ala-OH, N-Benzyl-N-Cbz-beta-alanine
A134695-10G
A134695-1G
A134695-25G
A134695-5G
76972-292EA
43.5
USD
76972-292
76972-294
76972-296
76972-298
N-Benzyl-N-Cbz-β-alanine 97%
N-Benzyl-N-Cbz-β-alanine
Precaución: for Research Use Only
Formula:
C₁₈H₁₉NO₄ MW: 313.348 g/mol |
MDL Number:
MFCD19703280 CAS Number: 252919-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...