2-(Furan-2-yl)-7-(2-(4-(4-(2-methoxyethoxy)phenyl)piperazin-1-yl)ethyl)-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine 98+%

Supplier: Ambeed
Warning

Synonyms: Preladenant

A120930-10MG A120930-1MG A120930-2MG A120930-50MG A120930-5MG A120930-100MG
76971-516EA 53.05 USD
76971-516 76971-518 76971-520 76971-522 76971-524 77566-240
2-(Furan-2-yl)-7-(2-(4-(4-(2-methoxyethoxy)phenyl)piperazin-1-yl)ethyl)-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine 98+%
2-(Furan-2-yl)-7-(2-(4-(4-(2-methoxyethoxy)phenyl)piperazin-1-yl)ethyl)-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine

Precaución: For research use only.

Formula: C₂₅H₂₉N₉O₃
MW: 503.56 g/mol
MDL Number: MFCD17167056
CAS Number: 377727-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR