8-Bromopyrido[4,3-d]pyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

A341621-100MG A341621-250MG
76978-422EA 47.75 USD
76978-422 77672-742
8-Bromopyrido[4,3-d]pyrimidin-4(3H)-one 97%
8-Bromopyrido[4,3-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O
MW: 226.03 g/mol
MDL Number: MFCD19689556
CAS Number: 1256353-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR