Ethyl-4-amino-6-chloronicotinate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-amino-6-chloronicotinate

A263383-1G A263383-250MG
76977-022EA 247.19 USD
76977-022 76977-024
Ethyl-4-amino-6-chloronicotinate 95%
Ethyl-4-amino-6-chloronicotinate

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MW: 200.63 g/mol
MDL Number: MFCD00834975
CAS Number: 380626-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR