2-(2-Aminoethyl)-1-methylpyrrolidine 97%
Supplier: Ambeed
|
Danger
|
A177309-10G
A177309-1G
A177309-25G
A177309-5G
76974-298EA
23
USD
76974-298
76974-300
76974-302
76974-304
2-(2-Aminoethyl)-1-methylpyrrolidine 97%
2-(2-Aminoethyl)-1-methylpyrrolidine
Precaución: for Research Use Only
Formula:
C₇H₁₆N₂ |
MDL Number:
MFCD00003175 CAS Number: 51387-90-7 UN: 2735 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...