3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 95%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-5-methylphenylboronic acid, pinacol ester

A192142-100MG A192142-1G A192142-250MG
76974-894EA 37.14 USD
76974-894 76974-896 76974-898
3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 95%
3-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₃
MW: 234.09 g/mol
MDL Number: MFCD12546605
CAS Number: 889659-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR