1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-1-yl)ethanone ≥95%

Supplier: Ambeed
Warning

A531290-100MG A531290-1G A531290-250MG
76980-094EA 40.32 USD
76980-094 76980-096 76980-098
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-1-yl)ethanone ≥95%
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₆H₂₂BNO₃
MW: 287.16 g/mol
MDL Number: MFCD11227113
CAS Number: 937591-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR