5-Bromo-4-chlorothieno[2,3-d]pyrimidine 95%
Supplier: Ambeed
|
Warning
|
A187961-100MG
A187961-1G
A187961-250MG
A187961-5G
76974-732EA
61.54
USD
76974-732
76974-734
76974-736
76974-738
5-Bromo-4-chlorothieno[2,3-d]pyrimidine 95%
5-Bromo-4-chlorothieno[2,3-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₂BrClN₂S MW: 249.5200 g/mol |
MDL Number:
MFCD11042739 CAS Number: 814918-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...