BI 6727 98%

Supplier: Ambeed
Warning

A232139-10MG A232139-50MG A232139-5MG A232139-100MG A232139-1MG A232139-1G A232139-250MG
76976-196EA 25 USD
76976-196 76976-198 76976-200 77612-270 77672-066 77881-600 77881-601
BI 6727 98%
BI 6727

Precaución: for Research Use Only

Formula: C₃₄H₅₀N₈O₃
MDL Number: MFCD20926414
CAS Number: 755038-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR