6-Aminoisoindolin-1-one 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-2,3-dihydro-1H-isoindol-1-one

A197166-100MG A197166-1G A197166-250MG A197166-5G
76975-096EA 19 USD
76975-096 76975-098 76975-100 77881-454
6-Aminoisoindolin-1-one 97%
6-Aminoisoindolin-1-one

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.17 g/mol
MDL Number: MFCD09701282
CAS Number: 675109-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR