ω-Chloro-3,4-difluoroacetophenone 98%

Supplier: Ambeed
Danger

A318737-25G A318737-5G
76978-032EA 26 USD
76978-032 77881-896
ω-Chloro-3,4-difluoroacetophenone 98%
ω-Chloro-3,4-difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O
MDL Number: MFCD03966888
CAS Number: 51336-95-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR