tert-Butyl spiro[indoline-3,4'-piperidine]-1'-carboxylate 95%

Supplier: Ambeed
Warning

A524818-1G A524818-250MG
76980-028EA 201.58 USD
76980-028 76980-030
tert-Butyl spiro[indoline-3,4'-piperidine]-1'-carboxylate 95%
tert-Butyl spiro[indoline-3,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₄N₂O₂
MDL Number: MFCD09953258
CAS Number: 180465-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR