4-Amino-1-ethyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

A733483-100MG A733483-5G A733483-250MG A733483-1G
76981-290EA 7 USD
76981-290 76981-292 77674-006 77882-613
4-Amino-1-ethyl-1H-pyrazole 98%
4-Amino-1-ethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₉N₃
MDL Number: MFCD05861667
CAS Number: 876343-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR