1-Boc-3-bromo-7-nitroindole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-7-nitroindole, N-BOC protected, 1-Boc-3-Bromo-7-nitroindole

A169948-10G A169948-1G A169948-250MG A169948-5G
76973-956EA 231.28 USD
76973-956 76973-958 76973-960 76973-962
1-Boc-3-bromo-7-nitroindole 97%
1-Boc-3-bromo-7-nitroindole

Precaución: for Research Use Only

Formula: C₁₃H₁₃BrN₂O₄
MW: 341.1600 g/mol
MDL Number: MFCD05864785
CAS Number: 914349-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR