Boc-O-methyl-L-serine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-2-amino-3-methoxypropionic acid, Boc-Ser(Me)-OH, N-tert-Butoxycarbonyl-O-methyl-L-serine, (2S)-2-Amino-3-methoxypropanoic acid, N-BOC protected

A158967-10G A158967-1G A158967-250MG A158967-25G A158967-5G
76973-438EA 19 USD
76973-438 76973-440 76973-442 76973-444 76973-446
Boc-O-methyl-L-serine 97%
Boc-O-methyl-L-serine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MW: 219.237 g/mol
MDL Number: MFCD00153314
CAS Number: 51293-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR