(R)-Propylene carbonate 95%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Methyl-1,3-dioxolan-2-one

A393142-100G A393142-10G A393142-25G A393142-5G
76978-974EA 18.04 USD
76978-974 76978-976 76978-978 76978-980
(R)-Propylene carbonate 95%
(R)-Propylene carbonate

Precaución: for Research Use Only

Formula: C₄H₆O₃
MW: 102.09 g/mol
MDL Number: MFCD00798265
CAS Number: 16606-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR