3-Bromopiperidin-2-one 95%

Supplier: Ambeed
Danger

A974804-100MG A974804-1G A974804-250MG
76982-044EA 151.71 USD
76982-044 76982-046 77674-430
3-Bromopiperidin-2-one 95%
3-Bromopiperidin-2-one

Precaución: for Research Use Only

Formula: C₅H₈BrNO
MW: 178.02 g/mol
MDL Number: MFCD09834399
CAS Number: 34433-86-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR