Boc-Arg(Z)-OH 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Arg(Cbz)-OH

A808986-1G A808986-25G A808986-5G
76981-682EA 42.44 USD
76981-682 76981-684 76981-686
Boc-Arg(Z)-OH 98%
Boc-Arg(Z)-OH

Precaución: for Research Use Only

Formula: C₁₉H₂₈N₄O₆
MW: 408.5 g/mol
MDL Number: MFCD00058482
CAS Number: 51219-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR