5-Fluoro-2-hydroxypyridine 97%

Supplier: Ambeed
Warning

A210047-100G A210047-10G A210047-1G A210047-250MG A210047-25G A210047-5G
76975-498EA 475.29 USD
76975-498 76975-500 76975-502 76975-504 76975-506 76975-508
5-Fluoro-2-hydroxypyridine 97%
5-Fluoro-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄FNO
MW: 113.09 g/mol
MDL Number: MFCD03092918
CAS Number: 51173-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR