1-Boc-1-methylhydrazine 97%

Supplier: Ambeed
Warning

Synonyms: 1-tert-Butoxycarbonyl-1-methylhydrazine, 2-Methylcarbazic acid tert-Butyl ester, 1-Boc-1-methylhydrazine, tert-Butyl 2-Methylcarbazate

A392212-100G A392212-10G A392212-1G A392212-25G A392212-500G A392212-5G
76978-954EA 116.7 USD
76978-954 76978-956 76978-958 76978-960 76978-962 76978-964
1-Boc-1-methylhydrazine 97%
1-Boc-1-methylhydrazine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂O₂
MW: 146.1876 g/mol
MDL Number: MFCD05669700
CAS Number: 21075-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR