5-Bromo-3-chloropyridin-2-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5-bromo-3-chloropyridine, 5-Bromo-3-chloro-2-pyridinamine
A110665-100MG
A110665-10G
A110665-1G
A110665-250MG
A110665-25G
A110665-100G
A110665-5G
76970-748EA
5
USD
76970-748
76970-750
76970-752
76970-754
76970-756
77570-598
77570-600
5-Bromo-3-chloropyridin-2-amine 98%
5-Bromo-3-chloropyridin-2-amine
Precaución: for Research Use Only
Formula:
C₅H₄BrClN₂ |
MDL Number:
MFCD11110443 CAS Number: 38185-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...