Methyl tetrahydro-2H-pyran-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A646096-100MG
A646096-1G
76980-778EA
9.55
USD
76980-778
77673-756
Methyl tetrahydro-2H-pyran-3-carboxylate 97%
Methyl tetrahydro-2H-pyran-3-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₁₂O₃ MW: 144.1684 g/mol |
MDL Number:
MFCD09839011 CAS Number: 18729-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...